Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELAVL4 Mouse, anti-Human, Unconjugated, Clone: 1B1F5, Proteintech
SDP

Catalog No. 502239882 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9882 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9882 Supplier Proteintech Group Inc Supplier No. 678351IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELAVL4, also named as HuD, is a 380 amino acid protein. The full name is neuronal embryonic lethal altered visual system-like(ELAVL) RNA-binding protein. ELAVL4 is expressed in brain and localized in the nucleus. PC12 cells expressing T7±HuD were treated with thio-specific (BMH) or amine-specific (DSS) chemical crosslinking reagents, and the resultant crosslinked complexes in the cell extracts were analyzed by immunoblotting with anti-T7 antibody. In addition to the monomeric HuD of 41 kDa, three specific crosslinked complexes could be detected using either of the crosslinkers. Two of the complexes had molecular masses of ∼80 and 120 kDa, and were likely to be the dimer and trimer of HuD, respectively. (PMID: 12384599 )

Specifications

Antigen ELAVL4
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 1B1F5
Concentration 500 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 1996
Target Species Human, Rat, Mouse, Rabbit, Pig
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.