Learn More
Electric Field Apparatus
Click to view available options
:
Electric Field Apparatus
Description
Includes
Electric field apparatus, Plastic petri dish on Acrylic base with feet and terminal pillars, bottle of Castor Oil, packet of Iron filings, two point charges, ringed terminals (small and large), two charged plates and instructions
Great for classroom use.
- Place on an overhead projector for viewing by an entire class
- Consists of plastic Petri dish on acrylic base with feet and terminal pillars
Specifications
Specifications
For Use With (Application) | Show the lines of an electric field produced by a large electrostatic charge |
For Use With (Equipment) | Overhead projector |
Includes | Electric field apparatus, Plastic petri dish on Acrylic base with feet and terminal pillars, bottle of Castor Oil, packet of Iron filings, two point charges, ringed terminals (small and large), two charged plates and instructions |
Product Type | Electric Field Apparatus |