Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Electrode Fill Solutions

Catalog No. p-7235899
Click to view available options
Electrode Fill Solution:
NA+/K+/CA++/CL-
pH
This item is not returnable. View return policy
This item is not returnable. View return policy

In vitro diagnostics material for use with RAPIDLab™ 1200 series electrodes.

TRUSTED_SUSTAINABILITY

Specifications

Electrode Fill Solution NA+/K+/CA++/CL-
For Use With (Equipment) RAPIDLab™ 1200 Series Electrodes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.