Learn More
Cytiva Electrode Holder for Electrophoresis Units
Catalog No. 45001598 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
GenePhor Electrophoresis System
Catalog No. For Use With (Equipment)
45-001-598 GenePhor Electrophoresis System
1 options
Catalog No. 45-001-598 Supplier Cytiva Supplier No. 80110655
Includes
Feet
For use in the Multiohor 2 gel e electrophoresis system to hold movable EPH/IEF electrodes during alignment of the ge
  • Provides uniform pressure over buffer strips or electrode strips during separation
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) GenePhor Electrophoresis System
Type Magnet