Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electrode Storage Solution, LabChem™

Catalog No. p-5379545
Click to view available options
Quantity:
500 mL
1 L
4 L

Specifications

Chemical Name or Material Electrode Storage Solution
Packaging Poly Bottle
CAS Max % 98.4, 1, 0.6
CAS 877-24-7, 7447-40-7
Identification Passes Test
Physical Form Liquid
Linear Formula KHC8H4O4
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 204.23
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.