Learn More
Cytiva Electrodes and Paper for NovaBlot™
Catalog No. 45001596 Shop All Cytiva Products
Change view
Click to view available options
Type:
Electrode Paper
Catalog No. Type
45-001-596 Electrode Paper
1 options
Catalog No. 45-001-596 Supplier Cytiva Supplier No. 80110619
These Electrodes and Paper for NovaBlot™ are accessory items for NovaBlot™ Kit, the Multiphor™ II Electrophoresis System.
  • For NovaBlot Kit
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) NovaBlot Kit
Type Electrode Paper
Quantity 500 Pk.
Dimensions (L x W) 200 x 250 mm