Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Electrophoresis Apparatus with 7 x 14 cm Casting Tray
SCI_ED

Catalog No. S139035 Shop All Walter Products Products
Change view
Click to view available options
For Use With (Equipment):
Power Supply and 500 mL Buffer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S139035 Power Supply and 500 mL Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139035 Supplier Walter Products™ Supplier No. EL100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apparatus for performing gel electrophoresis.

  • UV transparent chamber is made from thick acrylic, making it durable and leak-proof
  • Lid fits securely in place and can easily be removed
  • All electrophoresis units feature adjustable leveling feet, recessed electrical wires, and a safety stop that prevents the gel from running when the cover is not securely fitted
  • Each casting tray with embossed gel ruler has rubber end caps and multiple slots for easy and versatile comb placement
  • Requires 500 mL buffer
  • Kit includes injection molded UV transparent base, transparent cover, 7 x 14 cm casting tray with embossed gel ruler and rubber end caps, 8/10 comb combo, and two 6-tooth combs

Specifications

For Use With (Equipment) Power Supply and 500 mL Buffer
Type Electrophoresis Apparatus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.