Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Electrophoresis Apparatus with 7 x 7 cm Casting Tray
SCI_ED

Catalog No. S139036 Shop All Walter Products Products
Change view
Click to view available options
For Use With (Equipment):
Power Supply and 500 mL Buffer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S139036 Power Supply and 500 mL Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139036 Supplier Walter Products™ Supplier No. EL200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apparatus for performing gel electrophoresis.

  • Electrodes are locked together at power supply end to prevent incorrect placement
  • Chamber has two knobs at both ends that protrude through cover to assist in removal of lid
  • Raised tabs located on diagonal corners of casting tray allow for easy, safe insertion and removal from chamber
  • Casting trays have a gray strip to contrast between the wells and agarose to allow for easy dye loading
  • Casting trays are etched with a positive/negative sign at each end and have a frosted strip that matches up with the chamber to ensure correct placement
  • Cover must be placed correctly or it will not fit properly and unit will not work
  • Corrosion-resistant platinum wires are removable and can be replaced if needed
  • Requires 500 mL buffer
  • Inludes: Injection molded UV transparent base, transparent cover, 7 x 7 cm casting tray with embossed gel ruler and rubber end caps, 8/10 comb combo, two 6-tooth combs

Specifications

For Use With (Equipment) Power Supply and 500 mL Buffer
Type Electrophoresis Apparatus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.