Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Electrophoresis Power Supply
SCI_ED

Catalog No. S139038 Shop All Walter Products Products
Change view
Click to view available options
Max. Current:
250 mA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. Current
S139038 250 mA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139038 Supplier Walter Products™ Supplier No. ELPS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Power supply for use with electrophoresis apparatus.

  • Can be used on 75 V or 150 V with a maximum current of 250 mA
  • Run one or two cells simultaneously
  • Run time for gels: 40 to 50 min at 75 V; 20 to 30 min at 150 V

Specifications

For Use With (Application) Gel Electrophoresis
Max. Current 250 mA
Type Power Supply
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.