Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Electroporation Cuvettes Plus™

Catalog No. p-7222375
Click to view available options
Capacity (Metric):
90 μL
400 μL
800 μL
Width (Metric) Gap:
1 mm
2 mm
4 mm
Closure Color:
Blue
Gray
Yellow
Quantity:
10 Pk.
100/Pk., 25 Pk.
50 Pk.

Electroporation Cuvettes Plus™ for use in electroporation and electrofusion of bacteria, yeast, insect, plant and mammalian cells.

  • Cuvettes and safety stand protect both user and sample
  • Compatible with many commercially available electroporators
  • The cuvettes are molded with embedded polished aluminum electrodes, and gamma irradiated for guaranteed sterility
  • BTX™ cuvettes can obtain high field strengths up to 25.0 kV/cm
  • Three electrode gap sizes are available, 1 mm for bacteria and yeast, 2 mm for all cell types and 4 mm for mammalian cells
  • Round cuvette caps are leak resistant and allow for quick and easy one-finger removal
  • A 20-position cuvette rack is available - must be ordered separately
  • BTX™ disposable cuvettes Plus require no maintenance
    • Simply discard carefully after use

For 10 and 50 packs, the cuvettes are individually wrapped in a package that includes a disposable cuvette and a sterile transfer pipette, which allows for quick and easy removal of the sample after electroporation.

For bulk packages, the cuvettes are packaged in 24 bags containing 100 cuvettes per bag without transfer pipettes.

Specifications

Sterility Sterile
Type Electroporation Cuvette
Disposable Single-use
Capacity (Metric) 90 μL
Material Polished Aluminum
Closure Color Gray
Includes 10 x indivually wrapped cuvettes, Sterile pipet for removing the sample after electroporation
Quantity 10 Pk.
Width (Metric) Gap 1 mm
For Use With (Equipment) BTX Generators, BIORAD Gene Pulser and Micro Pulser Systems
For Use With (Application) Electroporation of Bactera and Yeast
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.