Learn More
GSC Go Science Crazy Electroscope with Metal Case and Indicator Gauge
Shop All GSC Products
Click to view available options
:
Electroscope Case-Gold Foil
Description
The unit is bigger than standard electroscopes for easy viewing. The leaf is larger and the wide radius of swing provides a visual and sensitive demonstration. Built-in scale helps to determine the amount of charge generated.
- Indicator gauge
- Large viewing area
- Highly sensitive to electrical charge
- The metal case is strong and resistant to breakage
- Instructions included
Specifications
Specifications
| Description | 50mL Capacity |
| Product Type | Electroscope |
| Material | Metal Case, Glass Front and Rear Windows |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov