Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Elephant Toothpaste
GREENER_CHOICESCI_ED

Catalog No. s42857 Shop All Aldon Products
Click to view available options
Class Size:
5 Lab Groups
CHOKING HAZARD - Small parts. Not for children under 3 yrs.
CHOKING HAZARD - Small parts. Not for children under 3 yrs.

A visually stimulating classroom demonstration. Innovating Science™ Elephant Toothpaste shows the effects of a catalyst on a chemical reaction.

  • Discussion topics can include basic chemical reactions, phase changes and catalysts

Specifications

Certifications/Compliance Product SDSs
Color White
Grade Level Elementary
Grade 2 to 5
Age 10 to 13
For Use With (Application) Science Education
Includes 1 Clear bottle, 5 grams of Yeast, 150 mL Hydrogen peroxide, 3% 30 mL Dish soap, Five 5 oz. Plastic cups, 5 Stir sticks
Quantity 1
Weight (English) 1 lb.
Dimensions (L x W x H) 10 x 7 x 5 in.
Class Size 5 Lab Groups
Product Type General Chemistry
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.