Learn More
Stoelting™ Elevated Plus Maze
SDP
Stoelting™ Elevated Plus-Maze is used to assess anxiety-like behavior in laboratory animals.
$3840.00 - $4140.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 2
Catalog Number Mfr. No. Animal Price Quantity & Availability  
Catalog Number Mfr. No. Animal Price Quantity & Availability  
10-000-302
SDP
View Documents
Stoelting™
60140
Mouse
Each for $3,840.00
Only null left
Add to Cart
10-000-323
SDP
View Documents
Stoelting™
60240
Rat
Each for $4,140.00
Only null left
Add to Cart
Description
Description
  • Maze exploits the conflict between the innate fear that rodents have of open areas versus their desire to explore novel environments
  • Open Arms are equipped with a height-adjustable lip to minimize falls
  • Lip or ledge can be adjusted from 0 to 15 mm to minimize falls and to provide optional thigmotactic cues
  • Available for rat or mouse
  • Closed arm walls are held solidly in slotted base, but lift off for easy cleaning
  • Grey, non reflective base plate
  • Removable walls and legs mean the maze can be stored flat when not in used
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.