Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518072
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518072 Supplier Invitrogen™ Supplier No. PA518072
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ELF1
Gene Accession No. P32519, Q60775
Gene Alias E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1
Gene Symbols ELF1
Host Species Goat
Immunogen Synthetic peptide sequence (AMKQNELLEPNSF) corresponding to the C-terminus amino acids of ELF1 (aa 607-619).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13709, 1997
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.