Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599241 Supplier Invitrogen™ Supplier No. PA599241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ELF1.

This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ELF1
Gene Accession No. P32519
Gene Alias E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1
Gene Symbols ELF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ELF1(Accession P32519), corresponding to amino acid residues I553-T603.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1997
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.