Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | ELF1 |
|---|---|
| Concentration | 0.46 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5943
![]()
|
Proteintech Group Inc
550291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to ELF1.
This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.Specifications
| ELF1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P32519 | |
| 1997 | |
| Peptide | |
| Primary | |
| -20°C | |
| ELF1 |
| 0.46 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E74 like factor 1, ELF1 | |
| ELF1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title