Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596697 Supplier Invitrogen™ Supplier No. PA596697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATSLHEGPT NQLDLLIRAV EASVHSSNAH CTDKTIEAAE ALLHMESPTC LRDSRSPVEV FVPPCVSTPE FIHAAMRPDV ITETVVEVST EESEPMDTSP IPTSPDSHEP MKKKKVGRKP KTQQSPISNG SPELGIKKKP; Positive Samples: HL-60, A-549, MCF7, Jurkat; Cellular Location: Nucleus.

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 36 kD subunit. This subunit can interact with the C-terminal region of PCNA. It forms a core complex with the 38 and 40 kDa subunits. The core complex possesses DNA-dependent ATPase activity, which was found to be stimulated by PCNA in an in vitro system. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELF2
Gene Accession No. Q15723, Q9JHC9
Gene Alias 2610036A20Rik; A230104O07Rik; AW111824; BB183398; E74 like ETS transcription factor 2; E74-like factor 2; E74-like factor 2 (ets domain transcription factor); E74-like factor 2 (ets domain transcription factor) isoform 1; E74-like factor 2 (ets domain transcription factor) isoform 2; elf2; ets domain transcription factor; ets family transcription factor ELF2A2; ets family transcription factor ELF2C; ETS-related transcription factor Elf-2; EU32; NERF; NERF-1A; NERF-1a,b; NERF-1B; NERF-2; new Ets-related factor
Gene Symbols ELF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ELF2 (NP_0068651).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1998, 69257
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.