Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elf4/MEF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Elf4/MEF |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Elf4/MEF Polyclonal specifically detects Elf4/MEF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Elf4/MEF | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q99607 | |
2000 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SSRVSSRSAPQGKGSSSWEKPKIQHVGLQPSASLELGPSLDEEIPTTSTMLVSPAEGQVKLTKAVSASSVPSNIHLGVA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Cancer | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
E74-like factor 4, E74-like factor 4 (ets domain transcription factor), ETS-related transcription factor Elf-4, MEFELFRMyeloid Elf-1-like factor | |
ELF4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title