Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF4 Mouse anti-Human, Clone: OTI4D5, lyophilized, TrueMAB™
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF810142

Encompass_Preferred

Catalog No. 50-166-9910


May include imposed supplier surcharges.
Only null left
Add to cart

Description

Description

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5′-WGGA-3′. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
Specifications

Specifications

ELF4
Monoclonal
Unconjugated
ELF4
ELFR, MEF
Mouse
Affinity Chromatography
RUO
2000
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Western Blot
OTI4D5
PBS with 8% trehalose and no preservative; pH 7.3
Q99607
ELF4
Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli.
100 μg
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.