Learn More
Invitrogen™ ELF4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ELF4.
Specifications
Specifications
| Antigen | ELF4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Elf4 |
| Gene Accession No. | Q99607, Q9Z2U4 |
| Gene Alias | AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743 |
| Gene Symbols | Elf4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.