Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111501 Supplier Invitrogen™ Supplier No. PA5111501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDDNEATSHT MSTAEVLLNM ESPSDILDEK QIFSTSEMLP DSDPAPAVTL PNYLFPASEP DALNRAGDTS DQEGHSLEEK ASREESAKKT Highest antigen sequence indentity to the following orthologs: Rat - 53%, Mouse - 52%.

ELF4 is transcriptional activator that binds to DNA sequences containing the consensus 5'-WGGA-3'. The protein acts synergistically with RUNX1 to transactivate the IL3 promoter. Also transactivates the PRF1 promoter in natural killer (NK) cells. This protein plays a role in the development and function of NK and NK T-cells and in innate immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF4
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Elf4
Gene Accession No. Q99607
Gene Alias AV314029; BC042423; E74 like ETS transcription factor 4; E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ELFR; ENSMUSG00000053512; ETS-related transcription factor Elf-4; Gm9907; MEF; myeloid elf-1 like factor; myeloid Elf-1-like factor; RGD1560743
Gene Symbols Elf4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ELF4. Recombinant protein control fragment (Product #RP-106996).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.