Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575263
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.
Specifications
ELF5 | |
Polyclonal | |
Unconjugated | |
ELF5 | |
E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13711, 2001, 366142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8VDK3, Q9UKW6 | |
ELF5 | |
Synthetic peptide corresponding to amino acids 182-230 of Human Elf-5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction