Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577158 Supplier Invitrogen™ Supplier No. PA577158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELF5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ELF5
Gene Accession No. Q8VDK3, Q9UKW6
Gene Alias E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5
Gene Symbols ELF5
Host Species Rabbit
Immunogen Recombinant full length Human ELF5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13711, 2001, 366142
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.