Learn More
Invitrogen™ ELF5 Polyclonal Antibody
Description
This antibody is predicted to react with Monkey, Rat, Rabbit.
Specifications
Specifications
| Antigen | ELF5 |
| Applications | ChIP Assay, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol, 0.1% BSA and 0.09% sodium azide; pH 7.4 |
| Gene | ELF5 |
| Gene Accession No. | Q8VDK3, Q9UKW6 |
| Gene Alias | E74 like ETS transcription factor 5; E74-like factor 5; E74-like factor 5 (ets domain transcription factor); E74-like factor 5 ESE-2a; Elf5; elf-5; Epithelium specific Ets-2; epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE2; ESE-2; ESE-5; ESE-5.; ets transcription factor; ETS-related transcription factor Elf-5 |
| Gene Symbols | ELF5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.