Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELF5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304468100UL
Description
ELF5 Polyclonal specifically detects ELF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ELF5 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
E74-like factor 5, E74-like factor 5 (ets domain transcription factor), Epithelium-restricted ESE-1-related Ets factor, Epithelium-specific Ets transcription factor 2, ESE2ESE-2, ETS-related transcription factor Elf-5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ELF5 (NP_001413.1). | |
100 μL | |
Cancer, Tumor Suppressors | |
2001 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction