Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELFN1 Antibody (965705), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10644PECY7
Description
ELFN1 Monoclonal antibody specifically detects ELFN1 in Human samples. It is validated for Flow CytometrySpecifications
ELFN1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
392617 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
965705 | |
Flow Cytometry | |
USP17L7 | |
Chinese Hamster Ovary cell line, CHO-derived human ELFN1 protein, Asp28-Tyr418, Accession # P0C7U0 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction