Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELFN1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA584721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584721 Supplier Thermo Scientific Supplier No. PA584721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSDMPCADDE CFSGDGTTPL VALPTLATQA EARPLIKVKQ LTQNSATITV QLPSPFHRMY TLEHFNNSKA STVSRLTKAQ EEIRLTNLF

ELFN1 Polyclonal antibody specifically detects ELFN1 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELFN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELFN1
Gene Accession No. P0C7U0
Gene Alias ELFN1; extracellular leucine rich repeat and fibronectin type III domain containing 1; extracellular leucine-rich repeat and fibronectin type III containing 1; extracellular leucine-rich repeat and fibronectin type III domain containing 1; extracellular leucine-rich repeat and fibronectin type-III domain-containing protein 1; PPP1R28; Protein ELFN1; protein phosphatase 1 regulatory subunit 28; protein phosphatase 1, regulatory subunit 28
Gene Symbols ELFN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELFN1
Purification Method Anitgen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 392617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.