Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELFN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551454 Supplier Invitrogen™ Supplier No. PA551454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIHHLEVKPA YHCSEHRHSF PALYYEEGAD SLSQRVSFLK PLTRSKRDST YSQLSPRHYY SGYSSSPEYS SESTHKIWER FRPYKKHHRE EVYMAAGHAL RKKVQFAKDE DLHDILDYWK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

ELFN2 is a single-pass membrane protein. It contains 1 fibronectin type-III domain and 5 LRR (leucine-rich) repeats. The exact function of ELFN2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELFN2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELFN2
Gene Accession No. Q5R3F8
Gene Alias 5leucine rich repeat containing 62; 6330514E13; AW048948; BC094219; dJ63G5.3 (putative Leucine rich protein); Elfn2; Elfn2-ps1; extracellular leucine rich repeat and fibronectin type III domain containing 2; extracellular leucine-rich repeat and fibronectin type III containing 2; extracellular leucine-rich repeat and fibronectin type III domain containing 2; extracellular leucine-rich repeat and fibronectin type III domain containing 2, pseudogene 1; extracellular leucine-rich repeat and fibronectin type III domain-containing protein 2; KIAA1904; leucine rich repeat and fibronectin type III, extracellular 2; leucine rich repeat containing 62; leucine-rich repeat and fibronectin type-III domain-containing protein 6; Leucine-rich repeat-containing protein 62; LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 29; Lrrc62; PPP1R29; protein phosphatase 1 regulatory subunit 29; protein phosphatase 1 regulatory subunit 29-like; protein phosphatase 1, regulatory subunit 29; RGD1559693
Gene Symbols ELFN2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELFN2. Recombinant protein control fragment (Product #RP-91477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114794
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.