Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELG-1Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991794
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1794 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1794 Supplier Rockland Immunochemicals Supplier No. 600401688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human Elg1 protein. A BLAST analysis of the immunogen suggest full to partial reactivity with several hypothetical proteins. This is not unexpected for such a novel protein. The following proteins should be considered to be highly homologous to Elg1 when using this antibody: human chromosome fragility associated gene 1 (NP_079133), hypothetical dog protein FLJ12735 (XP_548276), hypothetical chimpanzee protein FLJ12735 (XP_511388), and a hypothetical human protein of 1224 residues (CAH10412). All show 100% homology to the immunogen. Reactivity against homologues from other sources is not known.

Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.

Specifications

Antigen ELG-1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ATAD5
Gene Accession No. Q96QE3
Gene Alias ATAD5; ATPase family AAA domain-containing protein 5; ATPase family, AAA domain containing 5; C17orf41; chromosome fragility associated gene 1; Chromosome fragility-associated gene 1 protein; ELG1; enhanced level of genomic instability 1 homolog; FLJ12735; FRAG1
Gene Symbols ATAD5
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79915
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.