Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics MGB Alert™ C. pneumoniae and M. pneumoniae Real-Time PCR Assays

Catalog No. 22801073
Change view
Click to view available options
C. pneumonia Primer Mix
C. pneumonia Probe Mix
M. pneumonia Primer Mix
M. pneumonia Probe Mix
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-801-073 C. pneumonia Primer Mix
22-801-074 C. pneumonia Probe Mix
22-801-075 M. pneumonia Primer Mix
22-801-076 M. pneumonia Probe Mix
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-801-073 Supplier Nanogen, Inc Supplier No. 800433
This item is not returnable. View return policy

Specifications

Content And Storage One vial containing 30μL of chemically synthesized DNA Oligonucleotides
Kit Type C. pneumonia Primer Mix
No. of Tests 48
For Use With (Application) Pathogen Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.