Learn More
ELiTech Molecular Diagnostics MGB Alert™ Real-Time PCR Probes
Description
Proprietary MGB™ and Superbase™ technologies allow post-PCR dissociated melt curve analysis, providing molecular labs with more powerful tools for detecting nucleic acid sequences of specific organisms associated with disease. MGB Alert real-time PCR reagents are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into existing workflow. Laboratories may use MGB Alert reagents to develop real-time PCR assays.
- Compatible with most real-time PCR instruments
- Optimized for universal cycling conditions
- Easy to implement into existing workflow
- Improved analysis from post-PCR melt curve capabilities
MGB Alert BK Virus Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the VP1 region of BK Virus
MGB Alert CMV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from CMV
MGB Alert EBV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from EBV
MGB Alert Enterovirus Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from Enterovirus
MGB Alert HHV-6 Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HHV-6
MGB Alert hMPV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the Polymerase (L) gene of hMPV
MGB Alert HSV Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HSV
MGB Alert HSV 1 and 2 Probes consists of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from HSV 1 and 2
MGB Alert MS2 Probes and MS2-E Probes consist of a chemically synthesized DNA oligonucleotide probe that detects nucleic acid sequences from the MS2 bacteriophage
Specifications
Specifications
| Content And Storage | 300μL Chemically synthesized DNA oligonucleotide probe |
| For Use With (Application) | DNA Sequencing |
| No. of Reactions | 48 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.