Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELiTech Molecular Diagnostics Real-Time PCR Internal Controls

Catalog No. 22801072
Change view
Click to view available options
BK Virus Internal Controls
CMV 3.0 Internal Controls
EBV Internal Controls
HHV-6 Internal Controls
Internal Control Set IC1-V
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-801-072 Internal Control Set IC1-V
22-801-062 EBV Internal Controls
22-801-064 HHV-6 Internal Controls
22-801-065 BK Virus Internal Controls
22-801-066 CMV 3.0 Internal Controls
22-801-069 CMV 3.0 Internal Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-801-072 Supplier Nanogen, Inc Supplier No. 800439

missing translation for 'validMainframeMsg'

Available as FDA-cleared unassayed quality control materials, Real-Time PCR Internal Controls detect variation that may arise during the DNA or RNA extraction and amplification process associated with real-time PCR assays. Each set includes primers and probes that can be used to amplify and detect an internal control template (also included).

All Internal Controls use innovative MGB™ technology, including proprietary probe technology, providing labs with a powerful tool to detect variation in their real-time PCR assays. Real-Time PCR Internal Controls are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into an existing workflow.

  • Universal Cycling Conditions: Run multiple assays simultaneously
  • Reduced Royalties: Real-time PCR without TaqMan™ probes
  • Streamlined Workflow: Bring new molecular tests on quickly
  • Open Platform: Runs on most real-time PCR instruments

Specifications

Content And Storage Chemically synthesized DNA oligonucleotide primers and probes
Kit Type IC1-V Internal Control Set
No. of Tests 48
For Use With (Application) Real Time PCR (qPCR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.