Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Elk-1 Antibody (PCRP-ELK1-1B9), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308885UV
Description
Elk-1 Monoclonal antibody specifically detects Elk-1 in Human samples. It is validated for ELISA, Immunoprecipitation, Protein ArraySpecifications
Elk-1 | |
Monoclonal | |
DyLight 350 | |
ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
Recombinant full-length human Elk-1 protein (Uniprot: P19419) | |
100 μg | |
Autophagy, Cancer, MAP Kinase Signaling, Neuroscience | |
2002 | |
Store at 4°C in the dark. | |
IgG2a |
ELISA, Immunoprecipitation, Protein Array | |
PCRP-ELK1-1B9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction