Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587504 Supplier Invitrogen™ Supplier No. PA587504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSPLEACLEA EEAGLPLQVI LTPPEAPNLK SEELNVEPGL GRALPPEVKV EGPKEELEVA GERGFVPETT KAEPEVPPQE GVPARLPAVV MDTAGQA Positive Samples: HeLa, Jurkat, HT-29, K-562, Mouse liver, Rat brain; Cellular Location: Nucleus.

ELK1 is a component of the ternary complex that binds the serum response element (SRE) and mediates gene activity in response to serum and growth factors. ELK1 is phosphorylated by MAP kinase pathways at a cluster of S/T motifs at its C-terminus. Phosphorylation at these sites, particularly Ser383, is critical for transcriptional activation by ELK1. ELK1 appears to be a direct target of activated MAP kinase. Biochemical studies indicate that ELK1 is a good substrate for MAP kinase, the kinetics of ELK1 phosphorylation and activation correlate with MAP kinase activity, and interfering mutants of MAP kinase block ELK1 activation in vivo. ELK1 is a nuclear target for the ras-raf-MAPK signaling cascade. Alternatively spliced transcript variants encoding the same protein have been found for ELK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELK1
Gene Accession No. A4GTP4, P19419, P41969
Gene Alias ELK 1; ELK1; Elk-1; ELK1, ETS transcription factor; ELK1, member of ETS oncogene family; ETS domain-containing protein Elk-1; ETS transcription factor ELK1; ETS-like gene 1; Oncogene Elk1; tyrosine kinase (ELK1) oncogene
Gene Symbols ELK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 201-297 of human ELK1 (NP_0052202).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13712, 2002, 314436
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.