Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELK3 Antibody (OTI1H3), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270643AF350
Description
ELK3 Monoclonal specifically detects ELK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ELK3 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ELK3 | |
Full length recombinant protein of human ELK3 (NP_005221) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ELK3, ETS-domain protein (SRF accessory protein 2), ERPETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, NETSRF accessory protein 2, SAP-2, SAP2ETS-related protein NET, Serum response factor accessory protein 2 | |
Mouse | |
0.1 mL | |
2004 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction