Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELK3 Antibody (OTI1H3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270643MFV450
Description
ELK3 Monoclonal antibody specifically detects ELK3 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
ELK3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
2004 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1H3 | |
Flow Cytometry | |
ELK3, ETS-domain protein (SRF accessory protein 2), ERPETS domain-containing protein Elk-3, ETS-domain protein, ETS-related protein ERP, NETSRF accessory protein 2, SAP-2, SAP2ETS-related protein NET, Serum response factor accessory protein 2 | |
Full length human recombinant protein of human ELK3(NP_005221) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction