Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELK3 Monoclonal Antibody (OTI3D9), TrueMAB™, OriGene
SDP

Catalog No. 502540811
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0811 30 μL
50-167-4455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0811 Supplier OriGene Supplier No. TA503599S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ELK3 Monoclonal antibody specifically detects ELK3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen ELK3
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3D9
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ERBB2
Gene Accession No. P04626
Gene Alias D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 676-1255 of human ERBB2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2064
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.