Learn More
ELK3 Mouse anti-Human, Clone: OTI4G9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503575

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be a negative regulator of transcription, but can activate transcription when coexpressed with Ras, Src or Mos. Forms a ternary complex with the serum response factor and the ETS and SRF motifs of the Fos serum response element.Specifications
ELK3 | |
Monoclonal | |
Unconjugated | |
ELK3 | |
ERP, NET, SAP-2, SAP2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2004 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P41970 | |
ELK3 | |
Full length human recombit protein of human ELK3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.