Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535173
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535173 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535173 Supplier Invitrogen™ Supplier No. PA535173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ELK3 Polyclonal Antibody for Western Blot

The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELK3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ELK3
Gene Accession No. P41970
Gene Alias D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183
Gene Symbols ELK3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 98-126 amino acids from the N-terminal region of human ELK3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2004
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.