Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540875 Supplier Invitrogen™ Supplier No. PA540875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PVAPLSPARL QGANTLFQFP SVLNSHGPFT LSGLDGPSTP GPFSPDLQKT Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

The ELK4 gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by ELK4 is phosphorylated by the kinases, MAPK1 and MAPK8. This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is phosphorylated by the kinases, MAPK1 and MAPK8. Several transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELK4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ELK4
Gene Accession No. P28324
Gene Alias 2310011G17Rik; A130026I01Rik; BB162516; ELK4; ELK4, ETS transcription factor; ELK4, ETS-domain protein (SRF accessory protein 1); ELK4, member of ETS oncogene family; ETS domain-containing protein Elk-4; ETS domain-containing protein Elk-4-like protein; ETS transcription factor ELK4; Sap1; SAP-1; serum response factor accessory protein 1; SRF accessory protein 1
Gene Symbols ELK4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ELK4 (aa 382-431).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.