Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555994 Supplier Invitrogen™ Supplier No. PA555994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPMTVGRIE GDCESLNFSE VSSSSKDVEN GGKDKPPQPG AKTSSRNDYI HSGLYSSFTL NSLNSSNVKL FKLIKTENPA EKLAEKKSPQ EPTPSVIKFV TTPSKKPPVE PVAATISIGP S Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

The ELK4 gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by ELK4 is phosphorylated by the kinases, MAPK1 and MAPK8. This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is phosphorylated by the kinases, MAPK1 and MAPK8. Several transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELK4
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELK4
Gene Accession No. P28324
Gene Alias 2310011G17Rik; A130026I01Rik; BB162516; ELK4; ELK4, ETS transcription factor; ELK4, ETS-domain protein (SRF accessory protein 1); ELK4, member of ETS oncogene family; ETS domain-containing protein Elk-4; ETS domain-containing protein Elk-4-like protein; ETS transcription factor ELK4; Sap1; SAP-1; serum response factor accessory protein 1; SRF accessory protein 1
Gene Symbols ELK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELK4. Recombinant protein control fragment (Product #RP-100728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.