Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELL Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081668R
Description
ELL Polyclonal antibody specifically detects ELL in Human samples. It is validated for Western BlotSpecifications
ELL | |
Polyclonal | |
Western Blot | |
C19orf17, chromosome 19 open reading frame 17, DKFZp434I1916, Eleven-nineteen lysine-rich leukemia protein, ELL gene (11-19 lysine-rich leukemia gene), ELL1, elongation factor RNA polymerase II, MEN, RNA polymerase II elongation factor ELL | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human eleven-nineteen lysine-rich leukemia using the numbering given in entry NP_006523.1 (GeneID 8178). | |
0.1 mL | |
Cancer | |
8178 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction