Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ELL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541513
Description
Peptide sequence: GLSCGRVSDG SKVSVFHVKL TDSALRAFES YRARQDSVSL RPSIRFQGSQ Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 92%; Human: 100%; Mouse: 92%; Rat: 92%.
Elongation factor component of the super elongation complex (SEC), a complex required to increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA. Elongation factor component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:22195968, PubMed:23932780). Specifically required for stimulating the elongation step of RNA polymerase II- and III-dependent snRNA gene transcription (PubMed:23932780). ELL also plays an early role before its assembly into in the SEC complex by stabilizing RNA polymerase II recruitment/initiation and entry into the pause site. Required to stabilize the pre-initiation complex and early elongation. [UniProt].
Specifications
ELL | |
Polyclonal | |
Unconjugated | |
Ell | |
C19orf17; Eleven-nineteen lysine-rich leukemia protein; Ell; ELL gene (11-19 lysine-rich leukemia gene); ELL1; elongation factor for RNA polymerase II; elongation factor RNA polymerase II; hypothetical protein LOC325221; MEN; PPP1R68; protein phosphatase 1, regulatory subunit 68; RNA polymerase II elongation factor ELL; RNA POLYMERASE II ELONGATION FACTOR ELL (ELEVEN-NINETEEN LYSINE-RICH LEUKEMIA PROTEIN); wu:fc60b11; zgc:63836 | |
Rabbit | |
Protein A | |
RUO | |
8178 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P55199 | |
Ell | |
Synthetic peptide directed towards the N-terminal of human ELL (aa 11-60). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction