Learn More
Invitrogen™ ELL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120451
Description
Positive test controls include: Mouse testis, Mouse liver. The target is usually found in the following locations: Cajal body, Nucleus, Nucleus speckle. Immunogen sequence: PAVNGKLGVP NGREALLPTP GPPASTDTLS SSTHLPPRLE PPRAHDPLAD VSNDLGHSGR DCEHGEAAAP APTVRLGLPL LTDCAQPSRP HGSPSRSKPK K.
Elongation factor component of the super elongation complex (SEC), a complex required to increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA. Elongation factor component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:22195968, PubMed:23932780). Specifically required for stimulating the elongation step of RNA polymerase II- and III-dependent snRNA gene transcription (PubMed:23932780). ELL also plays an early role before its assembly into in the SEC complex by stabilizing RNA polymerase II recruitment/initiation and entry into the pause site. Required to stabilize the pre-initiation complex and early elongation. [UniProt].
Specifications
| ELL | |
| Polyclonal | |
| Unconjugated | |
| Ell | |
| C19orf17; Eleven-nineteen lysine-rich leukemia protein; Ell; ELL gene (11-19 lysine-rich leukemia gene); ELL1; elongation factor for RNA polymerase II; elongation factor RNA polymerase II; hypothetical protein LOC325221; MEN; PPP1R68; protein phosphatase 1, regulatory subunit 68; RNA polymerase II elongation factor ELL; RNA POLYMERASE II ELONGATION FACTOR ELL (ELEVEN-NINETEEN LYSINE-RICH LEUKEMIA PROTEIN); wu:fc60b11; zgc:63836 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13716 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O08856 | |
| Ell | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human ELL (NP_006523.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.