Learn More
ELL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301644A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.Specifications
| ELL | |
| Polyclonal | |
| Unconjugated | |
| ELL | |
| C19orf17, ELL1, MEN, PPP1R68, ELL gene (11-19 lysine-rich leukemia gene), RNA polymerase II elongation factor ELL, eleven-nineteen lysine-rich leukemia protein, protein phosphatase 1, regulatory subunit 68 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8178 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P55199 | |
| ELL | |
| Between 150 and 200 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.