Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELL Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560894
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0894 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0894 Supplier Bethyl Laboratories, Inc Supplier No. A301645AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Elongation factor that can increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA.

Specifications

Antigen ELL
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ELL
Gene Accession No. P55199
Gene Alias C19orf17, ELL1, MEN, PPP1R68, ELL gene (11-19 lysine-rich leukemia gene), RNA polymerase II elongation factor ELL, eleven-nineteen lysine-rich leukemia protein, protein phosphatase 1, regulatory subunit 68
Gene Symbols ELL
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8178
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.