Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100683 Supplier Invitrogen™ Supplier No. PA5100683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ELL2.

SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ELL2
Gene Accession No. O00472, Q3UKU1
Gene Alias cb244; eleven-nineteen lysine-rich leukemia gene 2; ELL2; ELL-related RNA polymerase II, elongation factor; elongation factor for RNA polymerase II 2; elongation factor RNA for polymerase II 2; elongation factor RNA polymerase II 2; elongation factor, RNA polymerase II 2; elongation factor, RNA polymerase II, 2; fb78f03; im:3814960; RNA polymerase II elongation factor ELL2; RNA polymerase II elongation factor ELL2-like protein; sb:cb244; wu:fb78f03; wu:fb78f03 protein
Gene Symbols ELL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ELL2(Accession O00472), corresponding to amino acid residues G161-I211.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192657, 22936
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.