Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ELL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556015 Supplier Invitrogen™ Supplier No. PA556015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HRGYLRLPGP GWSCLFSFIV SQCCQEGAGG SLDLVCQRFL RSGPNSLHCL GSLRERLIIW AAMDSIPAPS SVQGHNLTED ARHPESWQNT GGYSEGDAVS QPQMALEEVS VSDPLASN Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 71%.

Enhancer-binding elongation factor that specifically binds enhancers in embryonic stem cells (ES cells), marks them, and is required for their future activation during stem cell specification. Does not only bind to enhancer regions of active genes, but also marks the enhancers that are in a poised or inactive state in ES cells and is required for establishing proper RNA polymerase II occupancy at developmentally regulated genes in a cohesin-dependent manner. Probably required for priming developmentally regulated genes for later recruitment of the super elongation complex (SEC), for transcriptional activation during differentiation. Required for recruitment of P-TEFb within SEC during differentiation. Probably preloaded on germ cell chromatin, suggesting that it may prime gene activation by marking enhancers as early as in the germ cells. Promoting epithelial-mesenchymal transition (EMT). Elongation factor component of the super elongation complex (SEC), a complex required to increase the catalytic rate of RNA polymerase II transcription by suppressing transient pausing by the polymerase at multiple sites along the DNA. Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:22195968).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELL3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELL3
Gene Accession No. Q9HB65
Gene Alias A930015D22Rik; Ell3; elongation factor for RNA polymerase II 3; elongation factor RNA polymerase II-like 3; RNA polymerase II elongation factor ELL3; similar to elongation factor RNA polymerase II-like 3
Gene Symbols ELL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELL3. Recombinant protein control fragment (Product #RP-89344).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.