Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl acrylate, 99.5%, stabilized
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -71°C |
| Density | 0.9200g/mL |
| Boiling Point | 99°C |
| Flash Point | 16°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.40% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4050 to 1.407 |
| Linear Formula | H2C=CHCO2C2H5 |
| Quantity | 25 mL |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. Harmful if swallowed. Harmful in contact with skin. Toxic if inhaled |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Gently wash with plenty of soap and water. Ground/Bond container and |
| EINECSNumber | 205-438-8 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction