Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ellagic acid hydrate, 97%, may cont. up to 12% water

Catalog No. p-7022805
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 476-66-4 | C14H6O8 | 302.19 g/mol

Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ellagic acid used as selective, ATP-competitive inhibitor of casein kinase 2. Exhibits antitumor activity and inhibits glutathione S-transferase. It is also used as a Topo I and II, FGR, GSK, and PKA inhibitor.Commonly occurring plant polyphenol, inhibitor of glutathione S-transferase. Used for the assay of factor XIIa in plasma.Contact activation in blood coagulation.

Solubility
Slightly soluble in water. Soluble in 1M sodium hydroxide (10 mg/ml), EtOH, and pyridine.

Notes
Air sensitive and light sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container. Store in dark and away form air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 476-66-4
Molecular Formula C14H6O8
Molecular Weight (g/mol) 302.19
MDL Number MFCD00006914
InChI Key AFSDNFLWKVMVRB-UHFFFAOYSA-N
Synonym ellagic acid, benzoaric acid, lagistase, eleagic acid, alizarine yellow, elagostasine, 2,3,7,8-tetrahydroxychromeno 5,4,3-cde chromene-5,10-dione, ellagic acid dihydrate, llagic acid, acide ellagique
PubChem CID 5281855
ChEBI CHEBI:4775
IUPAC Name 6,7,13,14-tetrahydroxy-2,9-dioxatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1(15),4(16),5,7,11,13-hexaene-3,10-dione
SMILES OC1=CC2=C3C(OC(=O)C4=CC(O)=C(O)C(OC2=O)=C34)=C1O

Specifications

Melting Point 450°C (decomposition)
Quantity 5 g
Beilstein 47549
Sensitivity Air and light sensitive
Merck Index 14,3547
Solubility Information Slightly soluble in water. Soluble in 1M sodium hydroxide (10mg/ml),EtOH,and pyridine.
Formula Weight 302.20 (Anhydrous)
Percent Purity 97%
Assay may cont. up to 12% water
Chemical Name or Material Ellagic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-508-3
RTECSNumber DJ2620000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.